Pure Cellulose Chromatography Paper
Actual product may vary
Fisherbrand
Pure Cellulose Chromatography Paper
Pure cellulose papers with smooth surfaces are tested to assure uniformity and reproducibility, and offer optimum separation and sample resolution
Have Questions?
Catalog NumberFilm ThicknessWet Strength
FIS05-714-40.35 mm35 cm
Catalog number FIS05-714-4
Price (MYR)
-
Film Thickness:
0.35 mm
Wet Strength:
35 cm
Pure cellulose papers with smooth surfaces are tested to assure uniformity and reproducibility, and offer optimum separation and sample resolution
  • Thin paper (05-714-1) is recommended for analytical separations
  • Thick paper (05-714-4) is recommended for electrophoresis and general chromatography
  • Both grades are carefully tested for spot formation, capillary action, water flowrate1, and adsorption speed
  • Supplied in 46 × 57cm (18 × 22 in.) sheets
Specifications
Dimensions (L x W)18 x 22 x 22 in. (46 x 57 cm)
Film Thickness0.35 mm
For Use With (Application)Electrophoresis and General Chromatography
Length (English)18 x 22 in.
MaterialCellulose
Wet Strength35 cm
Width (English)22 in.
Width (Metric)57 cm
Length (Metric)46 cm
Unit SizePack