Specifications
CalibrationTo contain
Certifications/ComplianceUSP, ASTM E 438, ASTM D2709
Capacity (Metric)100 mL
DisposableReusable
MaterialBorosilicate Glass
For Use WithUsed for the determination of small quantities of solids in large volumes of liquids
Stopper Number16
Tolerance0 to 0.2mL in ±0.01mL, 25mL in ±1mL, >25mL in ±2mL
ShapePear
Length (Metric)160 mm
Diameter (Metric)58 mm
Graduations0 to 0.2 mL in 0.01 mL increments, 25, 50, and 100 mL
Product TypeCentrifuge Tube
Max. RCF800 x g
Unit SizePack