Nalgene™ 500mL LLDPE Safety Bottle Carrier
Actual product may vary
Nalgene™ 500mL LLDPE Safety Bottle Carrier
Nalgene™ 500mL LLDPE Safety Bottle Carrier
Nalgene™ 500mL LLDPE Safety Bottle Carrier
Nalgene™ 500mL LLDPE Safety Bottle Carrier
Thermo Scientific
Nalgene™ 500mL LLDPE Safety Bottle Carrier
Secure bottles and reduce chance of bumping, cracking or chipping with this safety bottle carrier.
Have Questions?
Catalog NumberHolds
FIS03-439-31Bottles (500 mL)
Catalog number FIS03-439-31
Price (MYR)
1,317.00
Each
Holds:
Bottles (500 mL)
Price (MYR)
1,317.00
Each
Secure bottles and reduce chance of bumping, cracking, or chipping. Thermo Scientific™ Nalgene™ 500mL LLDPE Safety Bottle Carrier offers greater rigidity and impact resistance. The sturdy, wide handle is molded in one piece with the base allowing a comfortable grip, even when the carrier is full. This linear low density polyethylene carrier holds up to six standard 500mL reagent bottles in any profile.
  • Cover latches securely to base and allow carriers to stack
  • Dimensions: 14L x 10W x 11 in. H (35 x 25 x 28cm)
Please recycle the corrugate cartons and any LDPE bags your Nalgene products are delivered in.
Specifications
DescriptionCarrier Safety Bottle LLDPE 500mL
ColorYellow
HoldsBottles (500 mL)
MaterialLLDPE
Positions6
StackableYes
No. per Case6
No. per Pack1
Product TypeBottle Carrier
Unit SizeEach