▸
Pumps
▸
Pump Parts and Accessories
▸
Pump Plunger Assemblies
▸
Hand-Powered Blower
Actual product may vary
Fisherbrand
Hand-Powered Blower
Provides continuous stream of air at low pressure
Have Questions?
Catalog Number
For Use With
FIS02-680
Blowpipes
Catalog number
FIS02-680
Price (MYR)
203.00
Each
Add to cart
For Use With:
Blowpipes
Price (MYR)
203.00
Each
Add to cart
Provides continuous stream of air at low pressure
Consists of tube bulb and 20 in. (51cm) outlet tube
Net-covered second bulb serves as reservoir
For use with blowpipes and other small apparatus
Made of latex rubber
Specifications
For Use With
Blowpipes
Includes
Tube bulb and 20 in. outlet tube
Overall Length (English)
28 in.
Overall Length (Metric)
71.12 cm
Type
Blower