PYREX™ Low Form Griffin Beakers
Actual product may vary
Fisherbrand
PYREX™ Low Form Griffin Beakers
Offers optimum balance between thermal shock resistance and mechanical strength
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FIS02-540T4000 mL
FIS02-540C10 mL
FIS02-540J150 mL
FIS02-540M600 mL
FIS02-540R2000 mL
Catalog number FIS02-540T
Price (MYR)
710.00
Each
Capacity (Metric):
4000 mL
Price (MYR)
710.00
Each
With pouring spout and extra-large marking spot
Corning™ PYREX™ Low Form Griffin Beakers offer optimum balance between thermal shock resistance and mechanical strength.
With pouring spout and extra-large marking spot.
Beakers, with spout, manufactured with uniform wall thickness, offers optimum balance between thermal shock resistance and mechanical strength. For convenience, the 250 through 4000 mL beakers have a double graduated metric scale to indicate approximate content. All sizes have an extra large marking spot. The 10 mL size is not graduated.
Specifications
Capacity (English)135.25 oz.
Diameter (English) Outer6.29 in.
Diameter (Metric) Outer16 cm
Capacity (Metric)4000 mL
MaterialBorosilicate Glass
For Use With (Application)For Optimum Balance Between Thermal Shock Resistance and Mechanical Strength
Graduation Range500 to 3500 mL
Height (English)9.84 in.
Height (Metric)25 cm
IncludesSpout
Increments250 mL
Label TypeWhite Marking
Quantity1/Pk.
TypeBeaker
DisposableReusable
GraduatedYes
Certifications/ComplianceASTM E960
ScaleDual White Metric Scale
StyleGriffin
Unit SizeEach