Specifications
Accuracy±5%
Capacity (English)8.45 oz.
Capacity (Metric)250 mL
MaterialBorosilicate Glass
For Use With (Application)For Measuring and Mixing Liquids
Graduation Range25 to 200 mL
IncludesSpout
Increments25 mL
Label TypeMatte Finish Enamel Marking
Quantity12 Pk.
TypeBeaker
AutoclavableAutoclavable
DisposableReusable
GraduatedYes
Certifications/ComplianceASTM E960, Type I
ScaleDual White Scale
StyleGriffin Beaded Top
Unit SizePack