Inoculating Turntable
Actual product may vary
Troemner
Inoculating Turntable
Hand-operated inoculating turntables
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric) Inside
FIS02-216-37211.4 cm
FIS02-216-37315 cm
Catalog number FIS02-216-372
Price (MYR)
2,110.00
Each
Diameter (Metric) Inside:
11.4 cm
Price (MYR)
2,110.00
Each
Hand-operated inoculating turntables
Talboys hand-operated inoculating turntables produce almost concentric circles of bacterial colonies that are evenly distributed across Petri dishes. Cast iron turntables are coated with gray baked acrylic enamel.
The large turntable accommodates 100mm or 150mm Petri dishes. It has a height of only 32mm (1.26 in.), ideal for steadying a forearm on the work surface while plating. The large turntable contains two circular sections. The top section has a raised gripper lip as well as a recessed center disk covered with a non-skid rubber lining.
Specifications
Diameter (English) Inside4.49 in.
Diameter (Metric) Inside11.4 cm
MaterialStainless Steel
Product TypeInoculating Turntable
For Use With (Equipment)100 mm dia. Petri Dishes
IncludesTripod base and center disk covered with non-skid rubber lining
Unit SizeEach