CarePacs™
Actual product may vary
Mettler Toledo
CarePacs™
For routine testing of balances
Have Questions?
Catalog NumberMassClassificationProduct Type
FIS01-912-10210 g & 200 gASTM Class 1Small Weight Set
Catalog number FIS01-912-102
Price (MYR)
6,015.00
Each
Mass:
10 g & 200 g
Classification:
ASTM Class 1
Product Type:
Small Weight Set
Price (MYR)
6,015.00
Each
Perform testing by using only two test weights (included). Save time and money, and rely on accurate measuring results; a unique approach to early detection of out-of-process tolerances.
  • Convenient, cost-effective way to limit the risk of working outside of specified process tolerances
  • Tests are performed in the upper and lower weighing range, typically using just two weights SOP's from Mettler Toledo give clear guidance as to how to perform testing reliably
  • Weight class and nominal value of weights are defined according to characteristics of each balance model
  • Comes with ergonomic tweezers, weight forks, cleanroom-approved gloves and cleaning cloths
  • Includes two Mettler Toledo-recommended, balance model-specific test weights
Benefits of CarePacs
  • Ensures accurate and safe operation of balances
  • Lower cost for initial calibration and recalibration compared with traditional weight sets
  • Validation of process tolerances as low as 0.03%
  • Gap between calibrations performed by a qualified service technician is monitored with the correct weights
  • Option to add third weight between 1mg and 100g for customer-specific testing
Specifications
Product TypeSmall Weight Set
Certifications/ComplianceISO9001, ISO14001 environmental standard
ClassificationASTM Class 1
MaterialStainless Steel
Weight (Metric)10, 200 g
Mass10 g & 200 g
IncludesTwo test weights: 10 and 200g, ergonomic tweezers, weight forks, cleanroom approved gloves and cleaning cloths; Without certificate
Quantity2/Set
Packaging QuantityMultiple
Unit SizeEach