Specifications
Certifications/ComplianceASTM
Closure TypeNone
ColorOrange with Black Print
Dimensions (L x W)24 x 12 in.
Length (Metric)61 cm
Width (Metric)30 cm
AutoclavableAutoclavable
IncludesPrinted in English
Width (English)12 in.
Length (English)24 in.
Thickness38 micron
MaterialHDPE
Temperature (Metric)127°C
Unit SizePack