Enterprise Services
Vendor Managed Inventory Services
Service Support Plans
Instrument Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See all Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Service Support Plans
Instrument Services
New Lab Project Services
Documents and Certificates
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Chemicals
▸
Water
▸
Analytical Water
▸
Laboratory Grade Analytical Water
▸
Water, Ultra Trace Elemental Analysis Grade, Fisher Chemical™
Element Search
Structure Search
Fisher Chemical
Water, Ultra Trace Elemental Analysis Grade, Fisher Chemical™
Packaged in specially precleaned LDPE bottles to protect product integrity
Have Questions?
Catalog Number
Quantity
Packaging
FGCW9-1
1 L
LDPE Bottle
Catalog number
FGCW9-1
Price (MYR)
3,315.00
Each
Add to cart
Quantity:
1 L
Packaging:
LDPE Bottle
Request bulk or custom format
Price (MYR)
3,315.00
Each
Add to cart
Specifications
Chemical Name or Material
Water
pH
7
CAS
7732-18-5
ChemAlert Storage Symbol
Gray
Color
Colorless
View more
Evaluated for 65 metal impurities in ppt range
Also specified for ≤1ppb for chloride, phosphate and sulfate anion impurities
Packaged in precleaned LDPE bottles to maintain product integrity