Chromplete™ Acetonitrile, HPLC/GC, meets ACS/USP analytical requirements
Chromplete™ Acetonitrile, HPLC/GC, meets ACS/USP analytical requirements
Chromplete™ Acetonitrile, HPLC/GC, meets ACS/USP analytical requirements
Chromplete™ Acetonitrile, HPLC/GC, meets ACS/USP analytical requirements
Fisher Chemical
Chromplete™ Acetonitrile, HPLC/GC, meets ACS/USP analytical requirements
The Chromplete product line from Thermo Scientific includes reagents for common analyses and separations with the consistency and precision you expect. Chromplete Acetonitrile is suitable for HPLC, GC, SFC, TLC, ACS, Spectrophotometry, and meets the analytical requirements of USP. | CAS: 75-05-8 | C2H3N | 41.05 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FGCT0010110001 LAluminum Jug
Catalog number FGCT001011000
Price (MYR)
0.00
Each
Quantity:
1 L
Packaging:
Aluminum Jug
Request bulk or custom format
Price (MYR)
0.00
Each
Specifications
Chemical Name or MaterialAcetonitrile
Shelf Life1825 days
Melting Point-51°F
Absorbance<0.1 at 195 nm, <0.05 at 200 nm, <0.02 at 210 nm, <0.01 at 220 nm
CAS75-05-8
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Chromplete Acetonitrile meets the analytical requirements of USP and is suitable for use in:
  • HPLC coupled with UV, VIS, PDA, ECD, FL, and RI detectors
  • GC- FID, GC- ECD, GC-TCD
  • Pesticide Analysis by GC or HPLC
  • UV Spectrophotometry
  • Preparative chromatography
  • Supercritical Fluid and Thin Layer chromatography
  • Reactions/Analyses requiring ACS compliance
  • Extraction/Concentration Analyses
RUO – Research Use Only