Permount™ Mounting Medium
Actual product may vary
Fisher Chemical
Permount™ Mounting Medium
For both mounting and long-term storage of slides
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Volume (English)
FGCSP15-500500 mL16.9 oz.
FCMSP15-100100 mL3.38 oz.
FCMSP15-500500 mL16.9 oz.
Catalog number FGCSP15-500
Price (MYR)
633.00
Each
Volume (Metric):
500 mL
Volume (English):
16.9 oz.
Price (MYR)
633.00
Each
For both mounting and long-term storage of slides
  • Low viscosity
  • Gives a thinner adhesive layer for better optical quality
  • A refractive index near that of fixed protein helps keep image free of optical imperfections such as diffraction halos
  • Suitable for mounting cover slips to slides with thick or thin specimens
  • Won't shift the colors of biological dyes
  • Won't lose its cohesiveness, adhesive quality, or color with age
  • Preserves most biological dyes with little or no fading when slides are stored in darkness
Note: Highly flammable liquid and vapor. May be fatal if swallowed and enters airways. Causes skin irritation. Causes serious eye irritation.
Specifications
ApplicationsMounting and long-term storage of slides
Volume (English)16.9 oz.
Product TypeSlide Mounting Media
PackagingAmber Glass
Volume (Metric)500 mL
Unit SizeEach