Acetonitrile with 0.1% Formic Acid (v/v), Optima™ LC/MS Grade
Acetonitrile with 0.1% Formic Acid (v/v), Optima™ LC/MS Grade
Acetonitrile with 0.1% Formic Acid (v/v), Optima™ LC/MS Grade
Fisher Chemical
Acetonitrile with 0.1% Formic Acid (v/v), Optima™ LC/MS Grade
0.1% Formic Acid in Acetonitrile (v/v), C2H3N, CAS Number-75-05-8, 64-18-6, formisoton, formylic acid, collo-bueglatt, formira, aminic acid, bilorin, acide formique, myrmicyl, hydrogen carboxylic acid, methanoic acid, 2.5L | CAS: 75-05-8
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
Packaging:
Glass Bottle
Catalog number FGCLS120-212
Price (MYR)
991.00
Each
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
991.00
Each
Specifications
GradeLC-MS
Item Name0.1% Formic Acid in Acetonitrile (v/v)
Quantity2.5 L
FormLiquid
Reagent TypeSolvent Blends
View more
0.1% Formic Acid in Acetonitrile (v/v), C2H3N, CAS Number-75-05-8, 64-18-6, acide formique, aminic acid, bilorin, collo-bueglatt, formira, formisoton, formylic acid, hydrogen carboxylic acid, methanoic acid, myrmicyl, 1L
  • Ideal high purity mobile phase blend for LC/MS and UHPLC-UV applications
  • Pre-mixed for ease-of-use
  • Low metal ion content to ensure minimal metal contamination
  • Formic acid (FA) is a common mobile phase additive that provides protons for LC/MS analysis
  • FA is also known to improve the peak shapes of the resulting separation