▸
Water
▸
Chromatography Water
▸
MS Grade Chromatography Water
▸
Water with 0.1% Formic Acid (v/v), Optima™ LC/MS Grade
Element Search
Structure Search
Fisher Chemical
Water with 0.1% Formic Acid (v/v), Optima™ LC/MS Grade
Have Questions?
Change view
Quantity:
4 L
2.5 L
Packaging:
View all
Glass Bottle
Catalog number
FGCLS118-4
Price (MYR)
635.00
Each
Add to cart
Quantity:
4 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
635.00
Each
Add to cart
Specifications
Absorbance
0.01 AU max. at 254nm, 0.55 AU max. at 230nm, 0.85 AU max. at 220nm, 1.25 AU max. at 210nm
Chemical Name or Material
0.1% Formic Acid in Water (v/v)
Name Note
Solvent Blends
pH
2
CAS
7732-18-5
View more
Ideal high purity mobile phase blend for LC/MS and UHPLC-UV applications
Pre-mixed for ease-of-use
Low metal ion content to ensure minimal metal contamination
Formic acid (FA) is a common mobile phase additive that provides protons for LC/MS analysis
FA is also known to improve the peak shapes of the resulting separation