▸
Buffers and Standards
▸
General Chemistry Solutions
▸
Hydrogen Peroxide, 30% (Certified ACS), Thermo Scientific™
Element Search
Structure Search
Fisher Chemical
Hydrogen Peroxide, 30% (Certified ACS), Thermo Scientific™
Have Questions?
Change view
Quantity:
500 mL
4 L
Packaging:
Poly Bottle
Catalog number
FGCH325-500
Price (MYR)
374.00
Each
Add to cart
Quantity:
500 mL
Packaging:
Poly Bottle
Request bulk or custom format
Price (MYR)
374.00
Each
Add to cart
Specifications
Chemical Name or Material
Hydrogen Peroxide, 30%
Color
Colorless
CAS
7722-84-1
ChemAlert Storage Symbol
Yellow
Boiling Point
108°C
View more
Hydrogen Peroxide, 30% (Certified ACS)