Kanamycin Sulfate (White Powder), Fisher BioReagents
Actual product may vary
Kanamycin Sulfate (White Powder), Fisher BioReagents
Kanamycin Sulfate (White Powder), Fisher BioReagents
Fisher Chemical
Kanamycin Sulfate (White Powder), Fisher BioReagents
For selecting transformed cells containing kanamycin resistance gene | CAS: 25389-94-0 | C18H38N4O15S | 582.575 g/mol
Have Questions?
Catalog NumberQuantity
FGCBP906-55 g
Catalog number FGCBP906-5
Price (MYR)
461.00
Each
Quantity:
5 g
Price (MYR)
461.00
Each
For selecting transformed cells containing kanamycin resistance gene
Specifications
Chemical Name or MaterialKanamycin Sulfate
TypeKanamycin Sulfate
Physical FormSolid
CAS25389-94-0
ChemAlert Storage SymbolGray
pH6.5 to 8.5
Ignition Residue< 1.0%
Recommended StorageRT
PackagingPoly Bottle
Potency> 750μg/mg
ColorWhite
Quantity5 g
Molecular FormulaC18H38N4O15S
Merck Index15, 5329
InChI KeyOOYGSFOGFJDDHP-IZQIRFRQSA-N
SMILESC1C(C(C(C(C1N)OC2C(C(C(C(O2)CN)O)O)O)O)OC3C(C(C(C(O3)CO)O)N)O)N.OS(=O)(=O)O
IUPAC Name(2R,3S,4S,5S,6R)-2-(aminomethyl)-6-[(1R,2S,3S,4R,6S)-4,6-diamino-3-[(2S,3R,4S,5R,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-2-hydroxycyclohexyl]oxyoxane-3,4,5-triol;sulfuric acid
Molecular Weight (g/mol)582.575
PubChem CID134129479
Formula Weight582.58
Unit SizeEach