▸
Buffers and Standards
▸
Life Science Buffers
▸
PBS, Phosphate Buffered Saline, 10X Solution, Fisher BioReagents™
Actual product may vary
Fisher Chemical
PBS, Phosphate Buffered Saline, 10X Solution, Fisher BioReagents™
Commonly used buffer
Have Questions?
Change view
Catalog Number
Quantity
Packaging
FGCBP399-1
1L
Nalgene Poly Bottle
FBRBP399-1
1L
Nalgene Poly Bottle
FBRBP399-500
500mL
Poly Bottle
3 Options
Catalog number
FGCBP399-1
Price (MYR)
538.00
Each
Add to cart
Quantity:
1L
Packaging:
Nalgene Poly Bottle
Price (MYR)
538.00
Each
Add to cart
Commonly used buffer
A concentrated, standard phosphate buffer solution.
Specifications
Recommended Storage
Room temperature
Physical Form
Liquid
Quantity
1L
Packaging
Nalgene Poly Bottle