▸
Biochemical Reagents
▸
Amino Acids
▸
Glycine (White Crystals or Crystalline Powder), Fisher BioReagents™
Element Search
Structure Search
Fisher Chemical
Glycine (White Crystals or Crystalline Powder), Fisher BioReagents™
Have Questions?
Change view
Quantity:
500 g
1 kg
Packaging:
Poly Bottle
Catalog number
FGCBP381-500
Price (MYR)
480.00
Each
Add to cart
Quantity:
500 g
Packaging:
Poly Bottle
Request bulk or custom format
Price (MYR)
480.00
Each
Add to cart
Specifications
Chemical Name or Material
Glycine
Melting Point
182°C
Absorbance
0.1 AU max. at 280nm
Ammonium
0.02% max.
CAS
56-40-6
View more
Glycine (White Crystals or Crystalline Powder)
Glycine is used in the preparation of Tris–Glycine electrophoresis buffers and culture media.