2-Propanol, Molecular Biology Grade, Fisher BioReagents™
2-Propanol, Molecular Biology Grade, Fisher BioReagents™
2-Propanol, Molecular Biology Grade, Fisher BioReagents™
Fisher Chemical
2-Propanol, Molecular Biology Grade, Fisher BioReagents™
For nucleic acid precipitation | CAS: 67-63-0 | C3H8O | 60.096 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
500 mL
1 L
4 L
Catalog number FGCBP2618-212
Price (MYR)
617.00
Each
Quantity:
2.5 L
Request bulk or custom format
Price (MYR)
617.00
Each
Specifications
Chemical Name or MaterialIsopropanol
pH7
Melting Point-89.5°C
Absorbance0.015 AU max. at 254nm, 0.10 AU max. at 230nm, 0.20 AU max. at 220nm, 1.00 AU max. at 205nm
CAS67-63-0
View more
For nucleic acid precipitation
Ultrapure reagent that can be used in fundamental applications such as purification and precipitation of nucleic acids.