Polyethylene Glycol 8000 (PEG), Fisher BioReagents™
Polyethylene Glycol 8000 (PEG), Fisher BioReagents™
Polyethylene Glycol 8000 (PEG), Fisher BioReagents™
Fisher Chemical
Polyethylene Glycol 8000 (PEG), Fisher BioReagents™
Polyethylene Glycol 8000, HO-[CH2CH2O]n-H, CAS Number-25322-68-3, Polyethylene Oxide, Carbowax, PEG 8000, 1kg, Gray, White, 7000 to 9000, CAUTION!
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
100 g
Packaging:
View all
Poly Bottle
Catalog number FGCBP233-1
Price (MYR)
595.00
Each
Quantity:
1 kg
Packaging:
Poly Bottle
Request bulk or custom format
Price (MYR)
595.00
Each
Specifications
Chemical Name or MaterialPolyethylene Glycol 8000
ColorWhite
pH5 to 7
CAS25322-68-3
ChemAlert Storage SymbolGray
View more
Polyethylene Glycol 8000, HO-[CH2CH2O]n-H, CAS Number-25322-68-3, Polyethylene Oxide, Carbowax, PEG 8000, 1kg, Gray, White, 7000 to 9000, CAUTION!
PEG 8000 is suitable as a medium for the fusion of mammalian cells. It is also used to precipitate bacteriophage from lysed cell supernatants.