Phenol/Chloroform/Isoamyl Alcohol (25:24:1 Mixture, pH 6.7/8.0, Liq.), Fisher BioReagents™
Phenol/Chloroform/Isoamyl Alcohol (25:24:1 Mixture, pH 6.7/8.0, Liq.), Fisher BioReagents™
Phenol/Chloroform/Isoamyl Alcohol (25:24:1 Mixture, pH 6.7/8.0, Liq.), Fisher BioReagents™
Fisher Chemical
Phenol/Chloroform/Isoamyl Alcohol (25:24:1 Mixture, pH 6.7/8.0, Liq.), Fisher BioReagents™
For extraction of proteins from crude nucleic acid preparations
Have Questions?
Change viewbuttonViewtableView
Quantity:
400 mL
100 mL
Packaging:
Coated Amber Glass/Poison Pack
Catalog number FGCBP1752I-400
Price (MYR)
1,202.00
Each
Quantity:
400 mL
Packaging:
Coated Amber Glass/Poison Pack
Request bulk or custom format
Price (MYR)
1,202.00
Each
Specifications
Chemical Name or MaterialPhenol/Chloroform/Isoamyl Alcohol
ColorClear
CAS108-95-2
ChemAlert Storage SymbolWhite
Health Hazard 1Signal Word: DANGER!
View more
For extraction of proteins from crude nucleic acid preparations
This mixture is used for extracting protein from crude nucleic acid preparations. Solution comes with a bottle of tris that should be used to equilibrate the pH.