▸
Solvents
▸
Acetonitrile
▸
Acetonitrile, Optima UHPLC/MS Grade
Element Search
Structure Search
Fisher Chemical
Acetonitrile, Optima UHPLC/MS Grade
Have Questions?
Catalog number
FGCA956-1
Price (MYR)
1,072.00
Each
Add to cart
Request bulk or custom format
Price (MYR)
1,072.00
Each
Add to cart
Specifications
Melting Point
-46°C
CAS
75-05-8
Color
Colorless
Quantity
1 L
Physical Form
Liquid
View more