Check Order Status
Hydrochloric Acid (TraceMetal™ Grade), Fisher Chemical™
Element Search
Structure Search
Fisher Chemical
Hydrochloric Acid (TraceMetal™ Grade), Fisher Chemical™
Have Questions?
Change view
Quantity:
2.5 L
Packaging:
HDPE Bottle
Catalog number
FGCA508-P212
Price (MYR)
1,016.00
EA
Add to cart
Quantity:
2.5 L
Packaging:
HDPE Bottle
Request bulk or custom format
Price (MYR)
1,016.00
EA
Add to cart
Specifications
Chemical Name or Material
Hydrochloric Acid
Shelf Life
3 years
pH
1
Cleanup Symbol
Acid
Color
Colorless
View more