Methanol, UHPLC-MS
Actual product may vary
Methanol, UHPLC-MS
Methanol, UHPLC-MS
Methanol, UHPLC-MS
Methanol, UHPLC-MS
Fisher Chemical
Methanol, UHPLC-MS
Unique signal-to-noise specification for the chemical industry where the solvent quality is linked directly to the sensitivity of the mass spectrometer. | CAS: 67-56-1 | CH4O | 32.04 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FGCA458-11 LClear Borosilicate Glass Bottle
Catalog number FGCA458-1
Price (MYR)
1,020.00
Each
Quantity:
1 L
Packaging:
Clear Borosilicate Glass Bottle
Price (MYR)
1,020.00
Each
Methanol, CH4O, CAS Number-67-56-1, wood naphtha, pyroxylic spirit, methyl alcohol, methylol, columbian spirit, colonial spirit, wood spirit, wood alcohol, methyl hydroxide, carbinol, 1L
RUO – Research Use Only
Specifications
Chemical Name or MaterialMethanol
Melting Point-98°C
Absorbance≤0.005 AU at 280nm, ≤0.01 AU at 254nm, ≤0.1 AU at 230nm, ≤0.4 AU at 215nm, ≤000 5 AU at 260nm, ≤02. AU at 220nm, ≤05. AU at 210nm
CAS67-56-1
ColorColorless
Recommended StorageRoom temperature
Density0.791 g/cm3
Quantity1 L
PackagingClear Borosilicate Glass Bottle
Physical FormLiquid
Residue after Evaporation1ppm max.
Assay Percent Range99.9% minimum by GC
Titratable Acid0.0003meq/g max.
Molecular FormulaCH4O
Titratable Base0.0002meq/g max.
Linear FormulaCH3OH
Vapor Pressure128 hPa at 20°C
MDL NumberMFCD00004595
Viscosity0.55 cP at 20°C
Water0.02% Max.
Synonymmethyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
Boiling Point64.7°C
InChI KeyOKKJLVBELUTLKV-UHFFFAOYSA-N
SMILESCO
IUPAC Namemethanol
Molecular Weight (g/mol)32.04
PubChem CID887
ChEBICHEBI:17790
GradeUHPLC-MS
Unit SizeEach