Forma™ FDE Series Ultra-Low Temperature GP Freezers
Actual product may vary
Forma™ FDE Series Ultra-Low Temperature GP Freezers
Forma™ FDE Series Ultra-Low Temperature GP Freezers
Forma™ FDE Series Ultra-Low Temperature GP Freezers
Forma™ FDE Series Ultra-Low Temperature GP Freezers
Forma™ FDE Series Ultra-Low Temperature GP Freezers
Forma™ FDE Series Ultra-Low Temperature GP Freezers
Forma™ FDE Series Ultra-Low Temperature GP Freezers
Thermo Scientific™ Forma™ FDE Series ultra-low temperature freezers feature reduced energy usage, a simplified touchscreen interface, four capacities, and an ergonomic cabinet design.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberElectrical RequirementsCapacity (Metric)Certifications/Compliance
FDE40086FV230 V 50 Hz549 LCE
FDE50086FV230 V 50 Hz682 LCE
Catalog number FDE40086FV
HSN 84184090
Price (MYR)
-
Electrical Requirements:
230 V 50 Hz
Capacity (Metric):
549 L
Certifications/Compliance:
CE
Part of the Standard Performance (STP) family, Thermo Scientific™ Forma™ FDE Series ultra-low temperature freezers, powered by H-drive, feature four upright models, maximizing storage capacity from 30,000 up to 60,000 2 mL vials. The FDE series is a sustainable and reliable solution with ultimate sample security and operational savings for every lab.
Performance
  • Powered by H-drive's hydrocarbon technology, these systems save up to 25% in energy usage and heat rejection over conventional refrigerant ultra-low freezers1
  • Designed for daily usage, recovers cabinet temperature 20% faster after opening the door.2
  • A quiet sound output below 53 dBA, enables the freezer to come out of the hallway and into the lab
  • Meets ±10°C validation protocols at -80°C with strong peak variation and steady-state temperature variation
  • Natural hydrocarbon refrigerants reduce system operating stresses promoting outstanding reliability and worry-free sample protection
User Friendly
  • The H-drive Information Center (HIC) combines easy to read temperature with simple capacitive touch buttons in a 5.6 inch display
  • Four capacities (300, 400, 500 and 600 2-inch box) to accommodate a variety of storage needs and lab footprint specifications
  • All models contain four inner doors allowing for easy sample access without exposing greater freezer content to ambient conditions
  • Improved customer convenience with new ergonomically designed handle, high contrast eye-level display, and comfortable easy-access shelf locations.
  • Compatible with most racking systems including a full range of Thermo Scientific™ racks for boxes, microplates, Thermo Scientific™ Matrix™ and Nunc CryoBank™ tubes
  • Security features including alarm settings and padlock-compatible doors
Sustainability
  • Manufactured in an award-winning, zero-waste to landfill facility
  • Lower HVAC costs as a result of less heat emission into the environment
  • Natural refrigerants and water-blown foam insulation, making the Series compliant with the U.S. Environmental Protection Agency's Significant New Alternatives Policy (SNAP), the European Union's F-Gas and other sustainability standards
Footnotes:
  1. Internal data comparing 11.2 kWh/day to UXF600 = 14.9 kWh/day. Data on file.
  2. Typical freezer test data based on internal testing in ambient temperature at 20°C; Typical standard validation protocol of ±10°C.
Specifications
DescriptionForma FDE Series Ultra-Low Freezers, 400-Box Capacity, 230V, 50Hz
TypeUltra-Low Freezer, Upright
Capacity (English)19.4 cu. ft.
Capacity (Metric)549 L
Temperature Range-50°C to -86°C
Door StyleOuter (1), Inner (4)
No. of Doors1
Inner Doors4
InsulationVacuum Insulation Panels (VIP) + Water-Blown Polyurethane Foam (SNAP-compliant)
InteriorStainless Steel
Shelves3
Capacity (English) Shelf165 lb.
Capacity (Metric) Shelf75 kg
Certifications/ComplianceCE
StyleUpright
DisplayHIC 5.6 in. 7-segment display with capacitive touch buttons
Product LineForma
WarrantyWarranty varies by country, please contact us for details
DefrostManual
Dimensions (DxWxH) Exterior38.5 x 28.3 x 78 in. (97.7 x 71.9 x 198.1 cm)
Dimensions (D x W x H) Interior28.3 x 23.1 x 51.2 in. (71.9 x 58.8 x 130.1 cm)
Monitoring OptionsAlarm, Recorder
Setpoint SecurityYes
ConnectivityExternal Monitoring/Cloud Compatible; 4-20mA, RS485, Dry Contacts
Smart View CompatibilitySmart-Vue Compatible
Amperage5.3/10 A
Plug TypeEU
Voltage230 V
Electrical Requirements230 V 50 Hz
Frequency50 Hz
Vial Capacity40,000 x 2mL
CompressorH-drive
RefrigerantR290 (1st stage), R170 + R290 mix (2nd stage)
Shipping Weight (English)687 lb.
Shipping Weight (Metric)312 kg
Weight (English)595 lb.
Weight (Metric)270 kg
Holds2 in. Boxes (400)
Unit SizeEach