▸
Water
▸
Multicompendial Water
▸
Purified Water, USP, Fisher Chemical™
Element Search
Structure Search
Fisher Chemical
Purified Water, USP, Fisher Chemical™
Have Questions?
Catalog Number
Quantity
Packaging
FCMW12-4
4 L
Plastic Bottle
Catalog number
FCMW12-4
Price (MYR)
459.00
Each
Add to cart
Quantity:
4 L
Packaging:
Plastic Bottle
Request bulk or custom format
Price (MYR)
459.00
Each
Add to cart
Specifications
Chemical Name or Material
Purified Water
Shelf Life
5 Years
pH
7
CAS
7732-18-5
Color
Colorless
View more