▸
Bases
▸
Sodium Hydroxide
▸
Sodium Hydroxide Solution (1N/Certified), Fisher Chemical™
Element Search
Structure Search
Fisher Chemical
Sodium Hydroxide Solution (1N/Certified), Fisher Chemical™
Volumetric standard solution, titrated at 25°C to pH 8.6 (phenolphthalein endpoint) against NIST™ potassium acid phthalate
Have Questions?
Catalog Number
Quantity
Packaging
FCMSS266-1
1 L
Poly Bottle
Catalog number
FCMSS266-1
Price (MYR)
440.00
Each
Add to cart
Quantity:
1 L
Packaging:
Poly Bottle
Request bulk or custom format
Price (MYR)
440.00
Each
Add to cart
Specifications
Chemical Name or Material
Sodium Hydroxide Solution
pH
13.3
Cleanup Symbol
Caustic
Color
Colorless
Name Note
1N
View more