Hydrochloric Acid Solution, 0.01N (N/100) (Certified), Fisher Chemical™
Actual product may vary
Hydrochloric Acid Solution, 0.01N (N/100) (Certified), Fisher Chemical™
Hydrochloric Acid Solution, 0.01N (N/100) (Certified), Fisher Chemical™
Fisher Chemical
Hydrochloric Acid Solution, 0.01N (N/100) (Certified), Fisher Chemical™
Standardized at 25°C to pH 8.2 against sodium hydroxide that has been standardized against NIST™ potassium acid phthalate | CAS: 7647-01-0 | ClH | 36.46 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FCMSA62-11 LPoly Bottle
Catalog number FCMSA62-1
Price (MYR)
499.00
Each
Quantity:
1 L
Packaging:
Poly Bottle
Price (MYR)
499.00
Each
Standardized at 25°C to pH 8.2 against sodium hydroxide that has been standardized against NIST™ potassium acid phthalate
Specifications
Chemical Name or MaterialHydrochloric Acid Solution, 0.01N (N/100)
pH0.10
Cleanup SymbolAcid
CAS7647-01-0
ChemAlert Storage SymbolGray
Concentration0.0099 to 0.0101N
Health Hazard 1Signal Word: CAUTION!
Health Hazard 2CAUTION!
CAS7732-18-5
Health Hazard 3Emergency Overview
May cause eye, skin, and respiratory tract irritation . The toxicological properties have not been fully investigated.
NFPA
Health:1 Flammability:0 Instability:0
IdentificationPass Test
PackagingPoly Bottle
Traceability to NISTLOT 84L Potassium acid phthalate
Quantity1 L
Molecular FormulaClH
Physical FormLiquid
MDL NumberMFCD00011324 MFCD00792839
UN Number1789
Synonymhydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
InChI KeyVEXZGXHMUGYJMC-UHFFFAOYSA-N
SMILESCl
IUPAC Namehydrogen chloride
Molecular Weight (g/mol)36.46
PubChem CID313
ChEBICHEBI:17883
GradeCertified
Unit SizeEach