▸
Acids
▸
Inorganic Acids
▸
Hydrochloric Acid
▸
Hydrochloric Acid Solution, 0.1N (N/10) (Certified), Fisher Chemical™
Element Search
Structure Search
Fisher Chemical
Hydrochloric Acid Solution, 0.1N (N/10) (Certified), Fisher Chemical™
Standardized at 25°C to pH 8.2 against sodium hydroxide that has been standardized against NIST™ potassium acid phthalate
Have Questions?
Catalog Number
Quantity
Packaging
FCMSA54-1
1 L
Poly Bottle
Catalog number
FCMSA54-1
Price (MYR)
542.00
Each
Add to cart
Quantity:
1 L
Packaging:
Poly Bottle
Request bulk or custom format
Price (MYR)
542.00
Each
Add to cart
Specifications
Chemical Name or Material
Hydrochloric Acid Solution, 0.1N (N/10)
pH
0.10 (1N)
Cleanup Symbol
Acid
CAS
7647-01-0
ChemAlert Storage Symbol