▸
Acids
▸
Inorganic Acids
▸
Sulfuric Acid
▸
Sulfuric Acid (70%/Textile), Fisher Chemical
Element Search
Structure Search
Fisher Chemical
Sulfuric Acid (70%/Textile), Fisher Chemical
Have Questions?
Catalog Number
Quantity
Packaging
FCMSA170-4
4 L
Poly Bottle
Catalog number
FCMSA170-4
Price (MYR)
604.00
Each
Add to cart
Quantity:
4 L
Packaging:
Poly Bottle
Request bulk or custom format
Price (MYR)
604.00
Each
Add to cart
Specifications
Chemical Name or Material
Sulfuric Acid
Cleanup Symbol
Acid
Name Note
70%
CAS
7664-93-9
ChemAlert Storage Symbol