Silica Gel Desiccant (100-200 Mesh/923, Chromatographic Grade), Fisher Chemical™
Silica Gel Desiccant (100-200 Mesh/923, Chromatographic Grade), Fisher Chemical™
Silica Gel Desiccant (100-200 Mesh/923, Chromatographic Grade), Fisher Chemical™
Silica Gel Desiccant (100-200 Mesh/923, Chromatographic Grade), Fisher Chemical™
Silica Gel Desiccant (100-200 Mesh/923, Chromatographic Grade), Fisher Chemical™
Fisher Chemical
Silica Gel Desiccant (100-200 Mesh/923, Chromatographic Grade), Fisher Chemical™
Silica gel Grade 923 conforms to the specifications of ASTM D1319 and ASTM D2549.
Have Questions?
Catalog NumberQuantityPackaging
FCMS679-500500 gPoly Bottle
Catalog number FCMS679-500
Price (MYR)
2,970.00
EA
Add to cart
Quantity:
500 g
Packaging:
Poly Bottle
Request bulk or custom format
Price (MYR)
2,970.00
EA
Add to cart
Specifications
Boiling Point2230°C
Chemical Name or MaterialSilica Gel Desiccant
ColorWhite
Melting Point1710°C
Physical FormSolid
View more
  • Sold as a case of 3 boxes - each box contains 24 x 10 g pouches
  • Suitable for use in Fluorescent Indicator Adsorption (FIA) test
  • Conforms to the specifications of ASTM D1319 and ASTM D2549
  • Surface area within the range of 430 - 530 m2/g
  • This grade of silica gel is also available in other sizes:
    • 500 g (S679500)
    • 2.5 kg (S679212)
    • 25 g (S67925)