Enterprise Services
Vendor Managed Inventory Services
Service Support Plans
Instrument Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See All Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Service Support Plans
Instrument Services
New Lab Project Services
Documents and Certificates
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Personal Protective Equipment
▸
Eye Protection and Face Protection
▸
General Purpose Safety Glasses
▸
Uvex™ Astrospec 3000™ Slim Safety Glasses
Actual product may vary
Fisher Chemical
Uvex™ Astrospec 3000™ Slim Safety Glasses
Smaller frame profile fits employees with slender faces
Have Questions?
Catalog Number
Frame Color
Lens Coating
Lens Tint
FCMS271-1
Blue
Scratch-resistant
Gray
Catalog number
FCMS271-1
Price (MYR)
461.00
Each
Add to cart
Frame Color:
Blue
Lens Coating:
Scratch-resistant
Lens Tint:
Gray
Price (MYR)
461.00
Each
Add to cart
Smaller frame size with adjustable lens inclination and temple length provides a custom fit for slim faces
Polycarbonate browguard and sideshields offer added protection against UV, dust, and flying particles
uvextreme™ Anti-Fog Coating (XTR)
Permanently bonded to lens
Absorbs 99.9% of UV radiation
Anti-scratch and -static
Will not wear after repeated cleanings
ultra-dura™ Hard Coat (UD)
Permanently bonded to lens
Scratch- and chemical-resistant
Specifications
Lens Coating
Scratch-resistant
Certifications/Compliance
Meet ANSI Z87-2003 and CSA Z94.3 standards.
Lens Tint
Gray
Frame Color
Blue
Weight (Metric)
33 g