Enterprise Services
Vendor Managed Inventory Services
Service Support Plans
Instrument Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See all Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Service Support Plans
Instrument Services
New Lab Project Services
Documents and Certificates
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Biochemical Reagents
▸
Staining Reagents
▸
Stains and Dyes
▸
Phenolphthalein (Certified ACS), Fisher Chemical™
Element Search
Structure Search
Fisher Chemical
Phenolphthalein (Certified ACS), Fisher Chemical™
Have Questions?
Catalog Number
Quantity
Packaging
FCMP79-100
100 g
Glass Bottle
Catalog number
FCMP79-100
Price (MYR)
501.00
Each
Add to cart
Quantity:
100 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
501.00
Each
Add to cart
Specifications
Chemical Name or Material
Phenolphthalein
Melting Point
258°C
CAS
77-09-8
ChemAlert Storage Symbol
Gray
Clarity
Pass Test
View more