NMP, N-Methylpyrrolidone, GC Headspace Grade, Thermo Scientific™
NMP, N-Methylpyrrolidone, GC Headspace Grade, Thermo Scientific™
NMP, N-Methylpyrrolidone, GC Headspace Grade, Thermo Scientific™
NMP, N-Methylpyrrolidone, GC Headspace Grade, Thermo Scientific™
Fisher Chemical
NMP, N-Methylpyrrolidone, GC Headspace Grade, Thermo Scientific™
Processed to high purity for accurate and repeatable determination of trace levels of residual solvents
Have Questions?
Catalog NumberQuantityPackaging
FCMN140-11 LGlass Bottle
Catalog number FCMN140-1
Price (MYR)
2,965.00
Each
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
2,965.00
Each
Specifications
Chemical Name or MaterialNMP, N-Methylpyrrolidone
Absorbance0.100 AU Maximum (325nm), 0.030 AU Maximum (350nm), 0.010 AU Maximum (400nm)
CAS872-50-4
ColorColorless to Yellow
Recommended StorageRoom temperature, packaged under inert atmosphere.
View more
  • Every lot is tested to ensure interference free baselines for interference free determination of Class 1, 2, and 3 residual solvents
  • 1 ppm max. of Class 1 Solvents, 10 ppm max. of Class 2 solvents, and 50 ppm max of Class 3 residual solvents
  • 1 ppm max. of Class 1 Solvents, 10 ppm max. of Class 2 solvents, and 50 ppm max of Class 3 residual solvents
  • Processed for low water content to facilitate the extraction of organic volatile impurities, or VOC