Enterprise Services
Vendor Managed Inventory Services
Service Support Plans
Instrument Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See all Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Service Support Plans
Instrument Services
New Lab Project Services
Documents and Certificates
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Buffers and Standards
▸
General Chemistry Solutions
▸
Hydrogen Peroxide, 30% (Certified ACS), Thermo Scientific™
Element Search
Structure Search
Fisher Chemical
Hydrogen Peroxide, 30% (Certified ACS), Thermo Scientific™
CAS:
7722-84-1 |
Molecular Formula:
H2O2 |
Molecular Weight:
34.014 g/mol
Have Questions?
Change view
Quantity:
4 L
500 mL
Packaging:
Poly Bottle
Catalog number
FCMH325-4
Price (MYR)
751.00
Each
Add to cart
Quantity:
4 L
Packaging:
Poly Bottle
Request bulk or custom format
Price (MYR)
751.00
Each
Add to cart
Specifications
Chemical Name or Material
Hydrogen Peroxide, 30%
Color
Colorless
CAS
7722-84-1
ChemAlert Storage Symbol
Yellow
Boiling Point
108°C
View more
Hydrogen Peroxide, 30% (Certified ACS)