Polymer Embeddings
Polymer Embeddings
Actual product may vary
Polymer Embeddings
Polymer Embeddings
Fisher Chemical
Polymer Embeddings
Have Questions?
Catalog NumberColorLength (Metric)Width (Metric)
FCME478-1White40.5 mm28.5 mm
Catalog number FCME478-1
Price (MYR)
735.00
EA
Add to cart
Color:
White
Length (Metric):
40.5 mm
Width (Metric):
28.5 mm
Price (MYR)
735.00
EA
Add to cart
Specifications
Quantity500/Pk.
ColorWhite
Length (Metric)40.5 mm
Width (Metric)28.5 mm
MaterialPolymer
Unit SizeEA