▸
Solvents
▸
Methylene Chloride
▸
Methylene Chloride (Stabilized/Certified ACS), Fisher Chemical™
Element Search
Structure Search
Fisher Chemical
Methylene Chloride (Stabilized/Certified ACS), Fisher Chemical™
Stabilized with preservative. Meet or exceeds the latest ACS specifications. Actual lot of analysis on label
Have Questions?
Catalog Number
Quantity
Packaging
FCMD37-500
500 mL
Glass Bottle
Catalog number
FCMD37-500
Price (MYR)
445.00
Each
Add to cart
Quantity:
500 mL
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
445.00
Each
Add to cart
Specifications
Chemical Name or Material
Methylene Chloride
Cleanup Symbol
Solvent
Melting Point
-97°C
Name Note
Stabilized
CAS
75-09-2
View more