▸
Solvents
▸
Methylene Chloride
▸
Methylene Chloride (GC Resolv™), Fisher Chemical
Element Search
Structure Search
Fisher Chemical
Methylene Chloride (GC Resolv™), Fisher Chemical
CAS: 75-09-2 | CH2Cl2 | 84.93 g/mol
Have Questions?
Catalog Number
Quantity
Packaging
FCMD154-4
4 L
Glass Bottle
Catalog number
FCMD154-4
Price (MYR)
864.00
Each
Add to cart
Quantity:
4 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
864.00
Each
Add to cart
Specifications
Chemical Name or Material
Methylene Chloride
Cleanup Symbol
Solvent
Melting Point
-97°C
CAS
75-09-2
ChemAlert Storage Symbol
Blue
View more