Cello-Seal™ Grease
Cello-Seal™ Grease
Actual product may vary
Fisher Chemical
Cello-Seal™ Grease
Won't add to pressure in vacuum system
Have Questions?
Catalog NumberQuantity
FCMC601-1006 Pk.
Catalog number FCMC601-100
Price (MYR)
875.00
EA
Add to cart
Quantity:
6 Pk.
Price (MYR)
875.00
EA
Add to cart
  • Low vapor pressure lubricant with consistency maintains consistency of petrolatum at 100°C (212°F)
  • May be blended with Cello-Grease™ Lubricant for heavier consistency
  • 100g jar
Specifications
Product TypeGrease
FormatLiquid
For Use With (Application)Won't add to pressure in vacuum system
For Use With (Equipment)Cello-Grease™ Lubricant
Boiling Point250°C
Melting Point90°C to 130°C
ColorAmber
PackagingGlass Jar
Quantity6 Pk.
Volume (English)3.52 oz.
Unit SizeEA