Chloroform, Stabilized with Amylene, for HPLC, Fisher Chemical™
Chloroform, Stabilized with Amylene, for HPLC, Fisher Chemical™
Chloroform, Stabilized with Amylene, for HPLC, Fisher Chemical™
Chloroform, Stabilized with Amylene, for HPLC, Fisher Chemical™
Fisher Chemical
Chloroform, Stabilized with Amylene, for HPLC, Fisher Chemical™
High purity solvent suitable for HPLC, spectrophotometry, and pesticide residue analysis
Have Questions?
Change viewbuttonViewtableView
Quantity:
4 L
2.5 L
Packaging:
View all
Amber Soda Lime Glass
Catalog number FCMC297-4
Price (MYR)
1,314.00
Each
Quantity:
4 L
Packaging:
Amber Soda Lime Glass
Request bulk or custom format
Price (MYR)
1,314.00
Each
Specifications
Chemical Name or MaterialChloroform
Cleanup SymbolSolvent
Melting Point-63°C
Name NoteApprox. 50ppm Amylene as Preservative
Absorbance0.01 AU max. at 280nm, 0.15 AU max. at 254nm, 0.32 AU max. at 250nm, 1.00 AU max. at 245nm
View more
  • Contains approx. 50ppm amylene as preservative
  • Screened for fluorescing and UV-absorbing contaminants to reduce interferences for smoother baselines
  • Tested to ensure minimum halogenated residue levels (≤10 ng/L) to demonstrate suitability for pesticide analysis
  • Also meets ACS specifications
  • Intended for research use only