▸
Bases
▸
Ammonium Hydroxide
▸
Ammonium Hydroxide (26° Bé/NF/FCC), Fisher Chemical
Element Search
Structure Search
Fisher Chemical
Ammonium Hydroxide (26° Bé/NF/FCC), Fisher Chemical
Have Questions?
Catalog Number
Quantity
Packaging
FCMA667-212
2.5 L
Glass
Catalog number
FCMA667-212
Price (MYR)
844.00
Each
Add to cart
Quantity:
2.5 L
Packaging:
Glass
Request bulk or custom format
Price (MYR)
844.00
Each
Add to cart
Specifications
Chemical Name or Material
Ammonium Hydroxide
pH
12
Cleanup Symbol
Caustic
Color
Colorless
Boiling Point
38°C
View more