▸
Acids
▸
Organic Acids
▸
Acetic Acid
▸
Acetic Acid Glacial Min. 99%, Trace Metal™, for Trace Metal Analysis, Fisher Chemical™
Element Search
Structure Search
Fisher Chemical
Acetic Acid Glacial Min. 99%, Trace Metal™, for Trace Metal Analysis, Fisher Chemical™
Have Questions?
Change view
Quantity:
500 mL
2.5 L
Catalog number
FCMA507P500
Price (MYR)
809.00
Each
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (MYR)
809.00
Each
Add to cart
Specifications
pH
2.5
CAS
64-19-7
Melting Point
16°C
Boiling Point
117°C
Color
Colorless
View more