Hydrofluoric Acid OPTIMA, Fisher Chemical™
Actual product may vary
Hydrofluoric Acid OPTIMA, Fisher Chemical™
Hydrofluoric Acid OPTIMA, Fisher Chemical™
Fisher Chemical
Hydrofluoric Acid OPTIMA, Fisher Chemical™
Hydrofluoric Acid OPTIMA 1L ultra trace analysis grade (PPT) | CAS: 7664-39-3 | FH | 20.01 g/mol
Have Questions?
Catalog number FCMA463-1
Price (MYR)
9,454.00
Each
Price (MYR)
9,454.00
Each
Hydrofluoric Acid OPTIMA 1L ultra trace analysis grade (PPT)
  • Hydrofluoric Acid OPTIMA 1L ultra trace analysis grade (PPT) Plastic Bottle Cas No. 7664-39-3
Specifications
Chemical Name or MaterialHydrofluoric Acid
CAS7664-39-3
Melting Point-35°C
Boiling Point105°C
PackagingPLASTIC
Quantity1 L
Molecular FormulaFH
MDL NumberMFCD00011346
InChI KeyKRHYYFGTRYWZRS-UHFFFAOYSA-N
SMILESF
IUPAC Namehydrogen fluoride
Molecular Weight (g/mol)20.01
Unit SizeEach