Methanol (HPLC), Fisher Chemical™
Methanol (HPLC), Fisher Chemical™
Methanol (HPLC), Fisher Chemical™
Methanol (HPLC), Fisher Chemical™
Fisher Chemical
Methanol (HPLC), Fisher Chemical™
HPLC mobile phase for analytical and preparative separations
Have Questions?
Catalog NumberQuantityPackaging
FCMA452-44 LGlass Bottle
Catalog number FCMA452-4
Price (MYR)
552.00
Bottle
Quantity:
4 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
552.00
Bottle
Specifications
Chemical Name or MaterialMethanol
Cleanup SymbolSolvent
Melting Point-98°C
Absorbance0.025 AU max. at 254nm, 0.15 AU max. at 230nm, 0.30 AU max. at 220nm, 1.00 AU max. at 205nm
CAS67-56-1
View more