▸
Acids
▸
Organic Acids
▸
Acetic Acid
▸
Acetic Acid, Glacial (HPLC), Fisher Chemical™
Element Search
Structure Search
Fisher Chemical
Acetic Acid, Glacial (HPLC), Fisher Chemical™
C2H4O2, CAS Number-64-19-7, 1L, 117 deg.C, Colorless, 60.05g/mol, 16 deg.C, 36152, Plastic Coated Glass bottle, 2.5, Liquid, 2.1, 1.52kPa at 20 deg.C
Have Questions?
Change view
Quantity:
500 mL
1 L
Packaging:
Glass Bottle
Plastic Coated Glass bottle
Catalog number
FCMA35-500
Price (MYR)
550.00
Each
Add to cart
Quantity:
500 mL
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
550.00
Each
Add to cart
Specifications
Absorbance
0.02 AU max. at 350nm, 0.05 AU max. at 280nm, 1.0 AU max. at 255nm
Chemical Name or Material
Acetic Acid, Glacial
Cleanup Symbol
Acid
Name Note
17.4N
pH
2.5
View more
C2H4O2, CAS Number-64-19-7, 1L, 117 deg.C, Colorless, 60.05g/mol, 16 deg.C, 36152, Plastic Coated Glass bottle, 2.5, Liquid, 2.1, 1.52kPa at 20 deg.C