▸
Acids
▸
Inorganic Acids
▸
Fluoroboric Acid
▸
Fluoroboric Acid, 50% (Laboratory), Fisher Chemical
Element Search
Structure Search
Fisher Chemical
Fluoroboric Acid, 50% (Laboratory), Fisher Chemical
Have Questions?
Catalog Number
Quantity
Packaging
FCMA188-500
500 mL
Poly Bottle
Catalog number
FCMA188-500
Price (MYR)
1,223.00
Bottle
Add to cart
Quantity:
500 mL
Packaging:
Poly Bottle
Request bulk or custom format
Price (MYR)
1,223.00
Bottle
Add to cart
Specifications
Chemical Name or Material
Fluoroboric Acid
Color
Yellow
Name Note
50%
CAS
16872-11-0
ChemAlert Storage Symbol