Enterprise Services
Vendor Managed Inventory Services
Service Support Plans
Instrument Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See all Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Service Support Plans
Instrument Services
New Lab Project Services
Documents and Certificates
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Chemicals
▸
Acids
▸
Organic Acids
▸
Formic Acid
▸
Formic Acid, 99.0+%, Optima™ LC/MS Grade, Fisher Chemical™
Element Search
Structure Search
Fisher Chemical
Formic Acid, 99.0+%, Optima™ LC/MS Grade, Fisher Chemical™
Suitable as a mobile phase additive for LC/MS and UHPLC/MS
Have Questions?
Change view
Quantity:
50 mL
10 x 1 mL
Packaging:
View all
Nalgene™ Poly Bottle
Catalog number
FCMA117-50
Price (MYR)
814.00
Bottle
Add to cart
Quantity:
50 mL
Packaging:
Nalgene™ Poly Bottle
Request bulk or custom format
Price (MYR)
814.00
Bottle
Add to cart
Specifications
Absorbance
0.01 AU max. at 254nm, 0.55 AU max. at 230nm, 0.85 AU max. at 220nm, 1.25 AU max. at 210nm
Chemical Name or Material
Formic Acid
Cleanup Symbol
Acid
pH
2.1
CAS
64-18-6
View more