▸
Buffers and Standards
▸
Life Science Buffers
▸
HEPES Buffer, 1M Solution, pH 7.3 (Molecular Biology), Fisher BioReagents™
Element Search
Structure Search
Fisher Chemical
HEPES Buffer, 1M Solution, pH 7.3 (Molecular Biology), Fisher BioReagents™
Commonly used buffering agent
Have Questions?
Change view
Quantity:
100 mL
500 mL
1 L
Packaging:
View all
Poly Bottle
Catalog number
FBRBP299-100
Price (MYR)
577.00
Each
Add to cart
Quantity:
100 mL
Packaging:
Poly Bottle
Request bulk or custom format
Price (MYR)
577.00
Each
Add to cart
Chemical Identifiers
CAS
7365-45-9, 7732-18-5
Specifications
Absorbance
≤0.01(0.1M solution) at 260nm, 0.004 max. (0.1M solution) at 280nm
Chemical Name or Material
HEPES Buffer
Name Note
1M Solution, pH 7.3
CAS
7365-45-9
ChemAlert Storage Symbol
Gray
View more
Commonly used buffering agent
HEPES is commonly used as a buffering agent.