Vancomycin Hydrochloride, Fisher BioReagents
Vancomycin Hydrochloride, Fisher BioReagents
Vancomycin Hydrochloride, Fisher BioReagents
Fisher Chemical
Vancomycin Hydrochloride, Fisher BioReagents
Inhibits formation of peptidoglycan polymers of bacterial cell wall
Have Questions?
Catalog NumberQuantity
FBRBP2958-11 g
Catalog number FBRBP2958-1
Price (MYR)
1,912.00
Each
Quantity:
1 g
Request bulk or custom format
Price (MYR)
1,912.00
Each
Specifications
Chemical Name or MaterialVancomycin Hydrochloride
TypeVancomycin Hydrochloride
Physical FormPowder/Solid
CAS1404-93-9
pH2.5 to 4.5
View more