▸
Organic compounds
▸
Unclassified Organic Compounds
▸
Guanidine Thiocyanate (White Crystalline Powder), Fisher BioReagents™
Element Search
Structure Search
Fisher Chemical
Guanidine Thiocyanate (White Crystalline Powder), Fisher BioReagents™
For the isolation of intact RNA from cell extracts
Have Questions?
Change view
Quantity:
250 g
1 kg
Packaging:
View all
Glass Bottle
Catalog number
FBRBP221-250
Price (MYR)
577.00
Each
Add to cart
Quantity:
250 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
577.00
Each
Add to cart
Specifications
Chemical Name or Material
Guanidine Thiocyanate
CAS
593-84-0
ChemAlert Storage Symbol
Blue
Absorbance
0.8 max. (70% in water at 25°C) at 280nm
Health Hazard 1
Signal Word: DANGER!
View more
For the isolation of intact RNA from cell extracts
A more powerful denaturing agent than Guanidine Hydrochloride, Guanidine Thiocyanate is suitable for the isolation of intact RNA from cell extracts.